Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, ≥99.0%, ReagentPlus™ Grade, Honeywell™

Catalog No. 60047900
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
Packaging:
Drum with liner
Fiberboard box
Poly bottle

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613

Specifications

Boiling Point 1.90°C (1.013hPa)
Melting Point 765°C
Color Colorless
pH 4,5 - 8
Physical Form Solid
Quantity 500 g
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
Hazard Statement P305 + P351 + P338
RTECSNumber 233-140-8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.