Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, ≥99.0%, ReagentPlus™ Grade, Solstice

Catalog No. 60047901 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
Packaging:
Drum with liner
Fiberboard box
Poly bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-901 5 kg Drum with liner
60-047-899 2.5 kg Poly bottle
60-047-900 500 g Poly bottle
60-047-898 1 kg Poly bottle
60-047-897 12 kg Fiberboard box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-047-901 Supplier Solstice Supplier No. C38815KG

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613

Specifications

Boiling Point 1.90°C (1.013hPa)
Melting Point 765°C
Color Colorless
pH 4,5 - 8
Physical Form Solid
Quantity 5 kg
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Drum with liner
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
Hazard Statement P305 + P351 + P338
RTECSNumber 233-140-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.