Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride dihydrate, 99% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 12312A3

Quantity 2kg

 View more versions of this product

Catalog No. AA12312A3


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10035-04-8
99% min.
147.02 (110.99Anhydrous)
Calcium chloride dihydrate
Granular
CaCl2·2H2O
MFCD00149613

Specifications

10035-04-8
99% min.
147.02 (110.99Anhydrous)
0.835g/mL
Calcium chloride dihydrate
Granular
CaCl2·2H2O
MFCD00149613
2kg

Safety and Handling

SDS & Safety Documents

EINECSNumber : 233-140-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit