Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, ACS, 99.0 to 102.0%, MilliporeSigma™

Catalog No. M1023821000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1023821000 1 kg Plastic Bottle
M1023820500 500 g Plastic Bottle
M1023825000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1023821000 Supplier MilliporeSigma Supplier No. 1023821000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·H2O
Synonym Calcium dichloride

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.0 to 102.0%
Linear Formula CaCl2·H2O
Formula Weight 147.2
Grade ACS/EP/Reagent
Packaging Plastic Bottle
Density 1.85g/cm3 (2°C)
Chemical Name or Material Calcium Chloride, Dihydrate, EMSURE™, For Analysis
Recommended Storage Store without Restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.