Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride dihydrate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99%, Solstice

Catalog No. 6002347 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-023-47 500 g
60-023-45 1 kg
60-023-46 2.5 kg
60-045-942 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-023-47 Supplier Solstice Supplier No. 31307500G

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O

Specifications

Quantity 500 g
MDL Number MFCD00149613
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Calcium Chloride Dihydrate, Reag. Ph. Eur.
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 600-075-5
RTECSNumber EV9810000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.