Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate Technical Flake, Reagents

Catalog No. 01337337
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-337 500 g
01-337-339 10 kg
01-337-338 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-337 Supplier Reagents Holdings Llc Supplier No. C2163700500A5
Add to Cart
Add to Cart

Calcium Chloride Dihydrate Technical Flake, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2H4O2
Molecular Weight (g/mol) 147.01
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
Synonym Calcium Dichloride Dihydrate
IUPAC Name calcium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ca++]

Specifications

Boiling Point >1600°C
Melting Point 176°C
Color Colorless/White
pH 4.5 to 8.5
Physical Form Solid
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Specific Gravity 1.835
Chemical Name or Material Calcium Chloride Dihydrate Technical Flake
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.