Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride hexahydrate, 98+%, for analysis, Acros Organics

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
7774-34-7
CaCl2·6H2O
219.08
MFCD00149616
QHFQAJHNDKBRBO-UHFFFAOYSA-L
calcium chloride hexahydrate,antarcticite,acmc-20alis,calcium hexahydrate dichloride,calcium chloride-water 1/6,calcium chloride hydrate, puratronic,calcium chloride, hydrated,calcium chloride hydrate trace metals basis 1g
6093252
calcium;dichloride;hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
7774-34-7
CaCl2·6H2O
219.08
MFCD00149616
QHFQAJHNDKBRBO-UHFFFAOYSA-L
calcium chloride hexahydrate,antarcticite,acmc-20alis,calcium hexahydrate dichloride,calcium chloride-water 1/6,calcium chloride hydrate, puratronic,calcium chloride, hydrated,calcium chloride hydrate trace metals basis 1g
6093252
calcium;dichloride;hexahydrate
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
Assay Percent Range 98% min. (Complexometry)
Linear Formula CaCl2·6H2O
Ammonium 0.01% max
Merck Index 15, 1661
Arsenic (As) 1ppm max.
Solubility Information Solubility in water: 5360g/L (20°C)
Formula Weight 219.08
Physical Form Chunks
Percent Purity 98+%
Grade Analysis
Copper (Cu) 0.002% max.
Insoluble in Water 0.025% max. (precipitated by NH4OH)
Insoluble Matter 0.025% max. (in water and ppt by NH4OH)
Iron (Fe) 0.005% max.
Lead (Pb) 0.002% max.
Nickel (Ni) 0.002% max.
Packaging Plastic bottle
Phosphate 0.003% max
Solubility Solubility in water: 5360g/L (20°C).
Sulfate 0.05% max
Color Colorless
Melting Point 30.0°C
pH 4.5 to 8.5 (5% soln.)
Quantity 2.5kg
Chemical Name or Material Calcium chloride hexahydrate, For analysis
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning