Learn More
Calcium Chloride Standard, 1000 ppm CaCO3 (400 ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 98.0, 0.19, 0.1 |
| CAS Max % | 100.0, 0.2, 0.1 |
| Equivalence | 1mL= 1 mg CaCO3 |
| Boiling Point | 100°C |
| Quantity | 4 L |
| Concentration or Composition (by Analyte or Components) | 1000ppm CaCO3 (400ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+ |
| Chemical Name or Material | Calcium Chloride Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.