Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calcium citrate tetrahydrate

Catalog No. 3597341KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 g
1 kg

Catalog No. 35-973-41KG

Supplier: Sigma Aldrich 3597341KG

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Thermal decomposition of calcium citrate tetrahydrate in dynamic air or dry nitrogen has been investigated by by tg, dta and dsc.calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition ca0.8 ba0.2 ti03 :pr3+, via sol-gel process
  • It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process
  • It is suitable for use in the preparation of bovine serum albumin (bsa) nanoparticles

Specifications

CAS 5785-44-4
Molecular Formula C12H10Ca3O14 -+ 4H2O
Linear Formula [O2CCH2C(OH)(CO2)CH2CO2]2 Ca3 -+ 4H2O
Quantity 1 kg
MDL Number MFCD00150786
Synonym Tricalcium dicitrate tetrahydrate
Molecular Weight (g/mol) 570.49
Percent Purity 99%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.