Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Citrate, Tetrahydrate, Powder, USP, 97.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. p-7157119 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail

C1102, 5785-44-4, C12H10Ca3O14·4H2O

Spectrum™ Chemical Calcium Citrate, Tetrahydrate, Powder, USP is used to treat conditions caused by low calcium levels such as bone loss (osteoporosis) and weak bones (osteomalacia/rickets). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5785-44-4
Molecular Formula C12H18Ca3O18
Molecular Weight (g/mol) 570.49
InChI Key LNIZKKFWMDARJV-UHFFFAOYSA-H
IUPAC Name tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
SMILES O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Loss on Drying 10.0-13.3%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 97.5-100.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.