Learn More
Calcium D-gluconate monohydrate, 98+%
Description
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
Solubility
Soluble in water(30g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Specifications
Specifications
Melting Point | ∼195°C (decomposition) |
Assay Percent Range | ≥98% |
Molecular Formula | C12H24CaO15 |
Quantity | 2500 g |
Merck Index | 14,1670 |
Synonym | calcium gluconate monohydrate, unii-czn0mi5r31, czn0mi5r31, d-gluconic acid calcium salt monohydrate, calcium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate, calcium d-gluconate monohydrate, calcium d-gluconate hydrate, d-gluconic acid calcium salt, calcium gluconate hydrate jan, calcicol tn |
Solubility Information | Soluble in water(30g/L). |
InChI Key | XLNFVCRGJZBQGX-XRDLMGPZSA-L |
SMILES | C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.O.[Ca+2] |
IUPAC Name | calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;hydrate |
Show More |
Safety and Handling
EINECSNumber | 206-075-8 |
RTECSNumber | EW2100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.