Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium D-gluconate monohydrate, 98+%

Catalog No. aaa1164936
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 66905-23-5 | C12H24CaO15 | 448.387 g/mol

Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.

Solubility
Soluble in water(30g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66905-23-5
MDL Number MFCD00064209

Specifications

Melting Point ∼195°C (decomposition)
Assay Percent Range ≥98%
Molecular Formula C12H24CaO15
Quantity 500 g
Merck Index 14,1670
Synonym calcium gluconate monohydrate, unii-czn0mi5r31, czn0mi5r31, d-gluconic acid calcium salt monohydrate, calcium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate, calcium d-gluconate monohydrate, calcium d-gluconate hydrate, d-gluconic acid calcium salt, calcium gluconate hydrate jan, calcicol tn
Solubility Information Soluble in water(30g/L).
InChI Key XLNFVCRGJZBQGX-XRDLMGPZSA-L
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.O.[Ca+2]
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;hydrate
Molecular Weight (g/mol) 448.387
PubChem CID 23724989
Formula Weight 448.40 (430.37 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Calcium D-gluconate monohydrate
Show More Show Less
EINECSNumber 206-075-8
RTECSNumber EW2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.