Learn More
Calcium fluoride, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Calcium fluoride is the commercially important source of fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers. It is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ca 2500°C |
Melting Point | 1403°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.5% (Metal basis) |
Beilstein | EM>5, |
Solubility Information | Slightly soluble in acid. Insoluble in acetone. |
IUPAC Name | calcium;difluoride |
Formula Weight | 78.08 |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335-H303 |
EINECSNumber | 232-188-7 |
RTECSNumber | EW1760000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.