Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Gluconate, Anhydrous, Powder, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601303 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-303 12 kg Fiber Drum
18-601-301 500 g Poly Bottle
18-601-302 2.5 kg Poly Pail
18-601-304 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-303 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C193912KG
Add to Cart
Add to Cart

C1939, 299-28-5, C12H22CaO14

Spectrum™ Chemical Calcium Gluconate, Anhydrous, Powder, USP is used as a nutrient. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 299-28-5
Molecular Formula C12H22CaO14
Molecular Weight (g/mol) 430.37
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
IUPAC Name calcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 1
Loss on Drying 0.03
Packaging Fiber Drum
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.