Calcium hydride, 90-95%, Alfa Aesar™

Manufacturer: Alfa Aesar 1910622

Quantity 100g

 View more versions of this product

Catalog No. AA1910622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-78-8
    Physical Form 2mm and down
    Assay Percent Range 90-95%
    Molecular Formula CaH2
    Formula Weight 42.1
    MDL Number MFCD00010897
    Chemical Name or Material Calcium hydride

    Specifications

    Formula Weight 42.1
    Quantity 100g
    EINECS Number 232-189-2
    Physical Form 2mm and down
    Assay Percent Range 90-95%
    Molecular Formula CaH2
    MDL Number MFCD00010897
    CAS 7789-78-8
    Chemical Name or Material Calcium hydride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit