Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 675.0°C |
| Quantity | 2.5 kg |
| Assay Percent Range | 92% min. (Total Ca) |
| Linear Formula | CaH2 |
| Fieser | 01,105 |
| Merck Index | 15,1674 |
| Solubility Information | Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts |
| Formula Weight | 42.09 |
| Percent Purity | ∼93% |
| Grade | Extra Pure |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement In contact with water releases flammable gases which may ignite spontaneously. Causes skin irritation. Causes serious eye irritation. Reacts violently with water. |
| missing translation for 'healthHazard3' | GHS P Statement Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages. IF ON S |
| EINECSNumber | 232-189-2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction