Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size

Catalog No. p-4833898
Click to view available options
Quantity:
100 g
500 g
2.5 kg

CAS: 7789-78-8 | CaH2 | 42.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897

Specifications

Melting Point 675.0°C
Quantity 100 g
Assay Percent Range 92% min. (Total Ca)
Linear Formula CaH2
Fieser 01,105
Merck Index 15,1674
Solubility Information Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Formula Weight 42.09
Percent Purity ∼93%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.9
Density 1.9000g/mL
Chemical Name or Material Calcium hydride, extra pure, 0-2 mm grain size
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
Causes serious eye irritation.
Reacts violently with water.
Precautionary Statement GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
IF ON S
EINECSNumber 232-189-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.