Learn More
Calcium hydrogen phosphate, anhydrous, 98% min
Description
It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | CaHPO4 |
Merck Index | 14,1692 |
Solubility Information | Soluble in hydrochloric acid,nitric acid,acetic acid,ammonium citrate solution. Slightly soluble in water. Insoluble in ethanol. |
Formula Weight | 136.06 |
Percent Purity | ≤98% |
Odor | Odorless |
Chemical Name or Material | Calcium hydrogen phosphate, Anhydrous |
Safety and Handling
EINECSNumber | 231-826-1 |
RTECSNumber | TB8528000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.