Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Hydroxide, Powder, EP, BP, JP, USP, 95-100.5%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
Description
Spectrum™ Chemical Calcium Hydroxide, Powder, USP, EP, BP, JP is used as an astringent that is topically applied in solutions or lotions. Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
Color | White |
CAS Max % | 100% |
Percent Purity | 95 to 100.5% |
Grade | Multi-Compendial |
Packaging | Poly Pail |
Quantity | 2.5 kg |
Chemical Name or Material | Calcium Hydroxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction