Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hypochlorite, Reagents

Catalog No. 01337332
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-332 500 g
01-337-333 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-332 Supplier Reagents Holdings Llc Supplier No. C2165000500A5
Add to Cart
Add to Cart

Calcium Hypochlorite, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
Molecular Weight (g/mol) 142.98
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym Bleaching powder, Chlorinated lime, Hypochlorous Acid calcium salt
IUPAC Name calcium dihypochlorite
SMILES [Ca++].[O-]Cl.[O-]Cl

Specifications

Melting Point 100°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Specific Gravity 2.35
Chemical Name or Material Calcium Hypochlorite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.