Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium hypochlorite, tech.

Catalog No. AA425480B
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA425480B 1 kg
AA4254818 50 g
AA4254830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA425480B Supplier Thermo Scientific Chemicals Supplier No. 425480B
Add to Cart

For Use in Deodorant, Oxidizing Agent, Bleaching Agent | CAS: 7778-54-3 | CaCl2O2 | 142.976 g/mol

Deodorant, oxidizing agent, bleaching agentCalcium hypochlorite is used for swimming pool sanitation and other water treatment plants. It is also used to disinfect drinking water, equipments in the industry and surfaces and equipments in kitchens. Further, it is used to bleach textiles and paper. It is also employed in bathroom cleaners and laundry detergents. It is used as an oxidizing agent in organic chemistry. It is involved in the cleavage of glycols, alfa-hydroxy carboxylic acids and keto acids to yield fragmented aldehydes or carboxylic acids respectively. It plays an important role in the haloform reaction to prepare chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-54-3
Molecular Formula CaCl2O2
MDL Number MFCD00010900
InChI Key ZKQDCIXGCQPQNV-UHFFFAOYSA-N
Synonym calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor
PubChem CID 24504

Specifications

Melting Point 100°C
Color White
Physical Form Granules
Quantity 1 kg
Solubility Information Soluble aq. soln..
SMILES [Ca++].[O-]Cl.[O-]Cl
IUPAC Name calcium dihypochlorite
Molecular Weight (g/mol) 142.98
Formula Weight 142.98
Appearance White granular powder
Density 2.35 g/cm3 (at 20°C)
Chemical Name or Material Calcium hypochlorite
Show More Show Less
Hazard Category H272-H290-H302-H314-H335
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378-P390-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 231-908-7
RTECSNumber NH3485000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.