Learn More
Calcium iodate monohydrate, 99+%
Description
Calcium iodate monohydrate is used as oxidant added to lotions and ointments, acts as antiseptic and deodorant. It is used in the manufacture of disinfectants and iodine supplement in animal feed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | >99% |
| Solubility Information | Soluble in nitric acid and slightly soluble in water. Insoluble in ethanol. |
| Formula Weight | 407.90 (389.88 Anhydrous) |
| Appearance | Off-white powder |
| Chemical Name or Material | Calcium iodate monohydrate |
Safety and Handling
| Hazard Category | H272-H315-H319-H335 |
| Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: II |
| EINECSNumber | 232-191-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.