Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium iodate, pure

Catalog No. AC318410025
Click to view available options
Quantity:
500 g
2.5 kg

Oxidation reagent | CAS: 7789-80-2 | CaI2O6 | 389.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-80-2
Molecular Formula CaI2O6
Molecular Weight (g/mol) 389.89
InChI Key UHWJJLGTKIWIJO-UHFFFAOYSA-L
Synonym calcium iodate, lautarite, calcium diiodate, unii-l8mn4y57br, l8mn4y57br, iodic acid hio3 , calcium salt 2:1, autarite, iodic acid, calcium salt, calcium iodate ca io3 2, hsdb 986
PubChem CID 24619
IUPAC Name calcium;diiodate
SMILES [O-]I(=O)=O.[O-]I(=O)=O.[Ca+2]

Specifications

Melting Point 540.0°C
Quantity 2.5 kg
Linear Formula Ca(IO3)2
Merck Index 15,1678
Solubility Information Solubility in water: 2g/L (15°C).
Formula Weight 389.89
Percent Purity ≥96% (Iodometry)
Grade Pure
Chemical Name or Material Calcium iodate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 232-191-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.