Alfa Aesar™ Calcium iodide hydrate, Puratronic™, 99.9965% (metals basis)

Manufacturer: Alfa Aesar™ 1111706

Quantity 5g

 View more versions of this product

Catalog No. AA1111706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71626-98-7
    Formula Weight 293.89 (Anhydrous)
    MDL Number MFCD00149622
    Merck Index 14,1677
    Chemical Name or Material Calcium iodide hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 293.89 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Merck Index 14,1677
    Color Yellow
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00149622
    Quantity 5g
    Solubility Information Freely soluble in water and alcohol
    Health Hazard 1 Warning
    Density 2.55g/mL
    Physical Form Crystalline
    CAS 71626-98-7
    Melting Point 42°C (dec.)
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material Calcium iodide hydrate
    Assay 99.9965%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see ... on this label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit