Calcium iodide, anhydrous, 99.5% (metals basis)

Manufacturer: Alfa Aesar 2239522

Quantity 100g

 View more versions of this product

Catalog No. AA2239522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-68-8
    Molecular Formula CaI2
    Formula Weight 293.89
    MDL Number MFCD00010910
    Merck Index 14,1677
    Chemical Name or Material Calcium iodide, Anhydrous

    Specifications

    Formula Weight 293.89
    Quantity 100g
    TSCA TSCA
    Merck Index 14,1677
    Solubility Information Freely soluble in water, methanol, ethanol, acetone; Insoluble in ether
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses,
    Physical Form Powder
    Assay Percent Range 99.5% (Metals basis)
    Molecular Formula CaI2
    MDL Number MFCD00010910
    CAS 10102-68-8
    Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
    Chemical Name or Material Calcium iodide, Anhydrous
    Assay 99.5%, (metals basis)

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses,

    H315-H319: Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit