Alfa Aesar™ Calcium iodide, ultra dry, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 4712122

Quantity 100g

 View more versions of this product

Catalog No. AA4712122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-68-8
    Formula Weight 293.89
    MDL Number MFCD00010910
    Merck Index 14,1677
    Chemical Name or Material Calcium iodide, Ultra dry

    Specifications

    Formula Weight 293.89
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    MDL Number MFCD00010910
    Boiling Point 1100°C
    Melting Point 784°C
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Mesh Size −10 Mesh
    Odor Odorless
    CAS 10102-68-8
    Color White to pale yellow
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Beads
    Sensitivity Light Sensitive & Hygroscopic
    Merck Index 14,1677
    Quantity 100g
    Storage Note 1
    • Ampouled under argon
    Density 3.956g/mL
    Chemical Name or Material Calcium iodide, Ultra dry
    Assay 99.995%, (metals basis)

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit