Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Lactate, Pentahydrate, FCC, EP, USP, 98-101%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Calcium Lactate, Pentahydrate, USP, FCC, EP is most commonly used as an antacid and to treat calcium deficiencies. Spectrum Chemical manufactured USP, FCC, EP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Loss on Drying | 22.0-27% |
| Packaging | Poly Pail |
| Quantity | 2.5 kg |
| Percent Purity | 98-101% |
| Grade | Multi Compendia |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction