Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis)

Catalog No. AA4451514
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 35054-52-5 | CaN2O6 | 164.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35054-52-5
Molecular Formula CaN2O6
Molecular Weight (g/mol) 164.09
MDL Number MFCD00149604
InChI Key ZCCIPPOKBCJFDN-UHFFFAOYSA-N
Synonym calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci
PubChem CID 16211464
SMILES [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 132°C (decomposition)
Melting Point 42°C
Color White
Physical Form Crystalline
Quantity 25 g
Linear Formula Ca(NO3)2·xH2O
UN Number UN1454
Merck Index 14,1682
IUPAC Name calcium;dinitrate;hydrate
Formula Weight 164.09 (Anhydrous)
Percent Purity 99.9995%
Grade Puratronic™
Density 1.82 g/mL
Assay (metals basis)
Chemical Name or Material Calcium nitrate hydrate
Show More Show Less
Hazard Category H272-H302-H318
Hazard Statement GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
EINECSNumber 233-332-1
RTECSNumber EW3000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.