Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium nitrate tetrahydrate, ACS, 99.0-103.0%

Catalog No. AA1236436
Click to view available options
Quantity:
500 g
2 kg

Manufacturing of other nitrates, fertilizers, matches | CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol

Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132°C (decomposition)
Melting Point 42°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0-103.0%
UN Number UN1454
Merck Index 14,1682
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 236.15 (164.09 Anhydrous)
Percent Purity 99.0 to 103.0%
Grade ACS
Sensitivity Hygroscopic
Density 1.82
Chemical Name or Material Calcium nitrate, tetrahydrate
Show More Show Less
Hazard Category H302-H318
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
EINECSNumber 233-332-1
RTECSNumber EW3000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.