Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Nitrate Tetrahydrate, Solstice

Catalog No. 6002316 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-023-16 500 g
60-023-14 1 kg
60-023-17 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-023-16 Supplier Solstice Supplier No. 31218500G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, 99-103%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Quantity 500 g
Linear Formula Ca(NO3)2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 236.15g/mol
Percent Purity 99 to 103%
Grade ACS/puriss. p.a.
Chemical Name or Material Calcium Nitrate Tetrahydrate
Hazard Statement P280-P305 + P351 + P338
Precautionary Statement H302-H318
EINECSNumber 233-332-1
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.