Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Oxide, Honeywell Fluka™

Catalog No. p-7071775 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
6 x 1 kg

Puriss., meets analytical specification of FCC, 96-100.5% (ex ignited substance), powder (fine)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Physical Form Fine Powder
Quantity 1 kg
Linear Formula CaO
UN Number 1910
IUPAC Name calcium oxidandiide
Formula Weight 56.08g/mol
Percent Purity 96 to 100.5% (ex ignited substance)
Grade puriss.
Density 3.3g/mL at 25°C
Chemical Name or Material Calcium Oxide, Meets analytical specification of FCC
Hazard Statement P261-P280-P305 + P351 + P338
Precautionary Statement H315-H318-H335
EINECSNumber 215-138-9
RTECSNumber EW3100000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.