Learn More
Calcium oxide, Reagent Grade
Description
Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Boiling Point | 2850°C |
| Melting Point | 2572°C |
| Physical Form | Powder |
| Quantity | 500 g |
| Solubility Information | Soluble in water with generation of heat,and forming Ca(OH)2,in acids,glycerol. Insoluble in alcoholSoluble in water. Insoluble in methanol,diethyl ether and n-octanol. |
| Formula Weight | 56.08 |
| Grade | Reagent |
| Refractive Index | 1.83 |
| Sensitivity | Air and moisture sensitive |
| Density | 3.25-3.38 |
| Show More |
Safety and Handling
| Hazard Category | H315-H318-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 215-138-9 |
| RTECSNumber | EW3100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.