Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium perchlorate tetrahydrate, pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
15627-86-8
CaCl2O8·4H2O
MFCD00191895
15627-86-8
CaCl2O8·4H2O
MFCD00191895
Oxidation reagent
Linear Formula CaCl2O8·4H2O
Formula Weight 311.04
Physical Form Crystals or Crystalline Powder
Percent Purity 12.2 to 13.5% (Calcium)
Grade Pure
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol
Color White
Quantity 100g
Chemical Name or Material Calcium perchlorate tetrahydrate

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger