Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Peroxide, FCC, 60%, Spectrum™ Chemical
SDP

Catalog No. 18601342 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
100 lb.
Packaging:
Amber Glass Bottle
Drum
Poly Pail

C1388, 1305-79-9, CaO2

Spectrum™ Chemical Calcium Peroxide, FCC is used as a food additive under the E number E930 as flour bleaching agent. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1305-79-9
Molecular Formula CaO2
Molecular Weight (g/mol) 72.08
InChI Key LHJQIRIGXXHNLA-UHFFFAOYSA-N
IUPAC Name calcium dioxidanediide
SMILES [Ca++].[O-][O-]

Specifications

CAS Max % 100%
UN Number UN1457
Quantity 125 g
Percent Purity 60%
Grade FCC
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.