Learn More
Calcium peroxide, typically 65%
Description
Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 500 g |
UN Number | UN1457 |
Merck Index | 14,1689 |
Solubility Information | Slightly soluble in water; Soluble in acids |
Formula Weight | 72.08 |
Percent Purity | 65% (typically) |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Calcium peroxide |
Safety and Handling
Hazard Category | H272-H314-H335 |
Hazard Statement | GHS H Statement H272-H314-H318 May intensify fire oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CALCIUM PEROXIDE |
EINECSNumber | 215-139-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.