Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Monobasic, MP Biomedicals™

Catalog No. MP219380380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219380380 100 g
MP219380350 50 g
MP219380390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219380380 Supplier MP Biomedicals, Inc Supplier No. 0219380380
Add to Cart
Add to Cart

Calcium Phosphate Monobasic, White Powder

  • It is mainly used in fertilizers, as acidulant in baking powders and in wheat flours, mineral supplement for foods and feeds, in enameling.
  • Phosphorus is an essential nutrient and therefore is a common component of agricultural fertilizers.
  • Calcium dihydrogen phosphate is used in the food industry as a leavening agent to cause baked goods to rise.
  • Because it is acidic, when combined with an alkali ingredient, commonly sodium bicarbonate (baking soda) or potassium bicarbonate, it reacts to produce carbon dioxide and a salt.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-23-8
Molecular Formula CaH4O8P2
Molecular Weight (g/mol) 234.05
MDL Number MFCD00010898
InChI Key YYRMJZQKEFZXMX-UHFFFAOYSA-L
Synonym Calcium dihydrogenphosphate, Acid calcium phosphate, Calcium biphosphate, Monocalcium orthophosphate

Specifications

Boiling Point Does not boil
Melting Point Loses water at 109°C; decomposes at 203°C (literature)
Color White
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in 1 M Hydrochloric acid (50mg/mL - clear, colorless solution), dilute HNO3, acetic acid; moderately soluble in water.
Vapor Pressure Zero
Density 2.22g/cm3 at 20°C(Lit.)
RTECSNumber TB8527000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.