Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate (pyro), 96% min

Catalog No. AA89836A7
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 7790-76-3 | Ca2O7P2 | 254.10 g/mol

Calcium phosphate is used in the production of phosphoric acid and fertilizers. It is used in baking as the acid in a leavening agent. It is also used in cheese products. This compound has also been used in gene transfection of cells. It is also being used in the development of contrast agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-76-3
Molecular Formula Ca2O7P2
Molecular Weight (g/mol) 254.10
MDL Number MFCD00015983
InChI Key JUNWLZAGQLJVLR-UHFFFAOYSA-J
Synonym calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2
PubChem CID 24632
ChEBI CHEBI:32598
IUPAC Name dicalcium;phosphonato phosphate
SMILES [Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O

Specifications

Melting Point 1230°C
Color White
Physical Form Powder
Quantity 5 kg
Solubility Information Soluble in hydrochloric acid and nitric acid. Insoluble in water.
Formula Weight 254.1
Percent Purity ≥96%
Odor Odorless
Density 3.09 g/mL
Chemical Name or Material Calcium phosphate (pyro)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-221-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.