Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium propionate, 98+%

Catalog No. p-3735913
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum

CAS: 4075-81-4 | C6H10CaO4 | 186.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4075-81-4
Molecular Formula C6H10CaO4
Molecular Weight (g/mol) 186.22
MDL Number MFCD00167354
InChI Key BCZXFFBUYPCTSJ-UHFFFAOYSA-L
Synonym calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907
PubChem CID 19999
ChEBI CHEBI:81716
IUPAC Name calcium;propanoate
SMILES [Ca++].CCC([O-])=O.CCC([O-])=O

Specifications

Melting Point >300.0°C
Color White
pH 6.9 to 9.0 (10% soln.)
Loss on Drying 4% max. (105°C, 2 Hrs)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (on dry substance) (Complexometry)
Packaging Plastic bottle
Linear Formula (CH3CH2COO)2Ca
Quantity 1 kg
Merck Index 15, 1700
Solubility Information Solubility in water: 260g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol,practically insoluble in acetone and benzene
Formula Weight 186.22
Percent Purity 98+%
Insoluble Matter 0.3% max. (in water)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Calcium propionate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-795-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.