Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium-sensing R/CaSR Antibody (HL 1499), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1001830AF594
Description
Calcium-sensing R/CaSR Monoclonal specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, ELISA.Specifications
Calcium-sensing R/CaSR | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CASR | |
N-terminal region of human Calcium Sensing Receptor. [UniProtNo. P41180] | |
Primary | |
Human, Mouse, Rat, Rabbit | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HL 1499 | |
Western Blot, ELISA | |
calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441 | |
Mouse | |
0.1 mL | |
846 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction