Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium-sensing R/CaSR Antibody (HL 1499) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 4 publications

$206.50 - $499.50

Specifications

Antigen Calcium-sensing R/CaSR
Clone HL 1499
Concentration 1.0 mg/mL
Dilution Western Blot 1:50 - 1:200, Simple Western 1:200, Flow Cytometry reported in scientific literature (PMID 33830943), ELISA 1:100 - 1:2000, Immunoprecipitation 5 - 10 ug, Radioimmunoassay
Applications Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001830SS
SDP
View Documents
Novus Biologicals
NB1001830SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB1001830
SDP
View Documents
Novus Biologicals
NB1001830
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Calcium-sensing R/CaSR Monoclonal specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunoprecipitation, Radioimmunoassay.
Specifications

Specifications

Calcium-sensing R/CaSR
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Immunofluorescence
Unconjugated
Mouse
Cytoskeleton Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction
P41180
846
N-terminal region of human Calcium Sensing Receptor. [UniProt# P41180]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
HL 1499
Western Blot 1:50 - 1:200, Simple Western 1:200, Flow Cytometry reported in scientific literature (PMID 33830943), ELISA 1:100 - 1:2000, Immunoprecipitation 5 - 10 ug, Radioimmunoassay
Monoclonal
Purified
RUO
Human, Mouse, Rat, Rabbit
calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
CASR
IgG1 κ
Protein G purified
130 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.