Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcium Sensing Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585137 Supplier Invitrogen™ Supplier No. PA585137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (94%), Dog (95%), Cat (95%), Pig (95%), Bovine (95%). Positive Control: 293T, A431, H1299, HeLa, mouse brain, mouse placenta. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcium Sensing Receptor
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CASR
Gene Accession No. P41180, Q9QY96
Gene Alias BoPCaR1; Ca sensing receptor; Ca(2+)-sensing receptor; calcium sensing receptor; calcium-sensing receptor; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; EIG8; Extracellular calcium-sensing receptor; FHH; FIH; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; Gprc2a; HHC; HHC1; HYPOC1; LOW QUALITY PROTEIN: extracellular calcium-sensing receptor; NSHPT; parathyroid Ca(2+)-sensing receptor 1; parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1
Gene Symbols CASR
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human Calcium Sensing Receptor. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12374, 846
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.