Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Calcium Sensor Dye eFluor™ 514

Catalog No. 5016959
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 50μg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-169-59 1mg
50-169-60 10 x 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-169-59 Supplier Invitrogen™ Supplier No. 65085939
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Membrane-permeable dye used to monitor changes in intracellular free calcium concentrations in the cell. Once eFluor™ 514 Calcium Sensor Dye enters the cell, cellular esterases cleave the AM group yielding a membrane-impermeable dye fluorescing at ≈520nm.

  • Calcium binding affinity: Kd = 232 nM
  • Molecular Weight: ≈1100 d
  • Peak Excitation: 490nm
  • Peak Emission: 514nm
  • Reconstitute in high-quality, freshly opened DMSO
  • Recommended concentration: 2 to 5 mM
  • Once reconstituted, protect from light and store dessicated at −20°C; avoid freeze-thawing
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
For Use With (Application) Calcium Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Product Line eBioscience
Quantity 1mg
Emission Wavelength 490/514

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.