Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Stearate, Thermo Scientific™

Click to view available options
Quantity:
3kg
66071-81-6
C36H70CaO4
607.03
MFCD00036390
CJZGTCYPCWQAJB-UHFFFAOYSA-L
calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates
15324
calcium;octadecanoate
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2]
66071-81-6
C36H70CaO4
607.03
MFCD00036390
CJZGTCYPCWQAJB-UHFFFAOYSA-L
calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates
15324
calcium;octadecanoate
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2]

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identification Pass Test
Linear Formula Ca(CH3(CH2)16COO)2
Quantity 3kg
Formula Weight 607.02
Chemical Name or Material Calcium stearate

RUO – Research Use Only