Learn More
Calcium sulfate, anhydrous, 99%
Description
Used as Portland-cement retarder; tile and plaster; It is the source of sulfur and sulfuric acid; Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1450°C |
Color | White |
Physical Form | Powder |
Quantity | 5 kg |
Assay Percent Range | 99% |
Solubility Information | Soluble in water,slightly soluble in glycerol. |
Formula Weight | 136.14 |
Sensitivity | Hygroscopic |
Appearance | White powder |
Density | 2.96 |
Show More |
Safety and Handling
Hazard Category | H315-H319 : Causes skin irritation. Causes serious eye irritation. |
EINECSNumber | 231-900-3 |
RTECSNumber | WS6920000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.