Learn More
Calcium Sulfate, Dihydrate, NF, 50 kg, Spectrum Chemical
SDP
Catalog No. 18615375
Click to view available options
:
Quantity: 50 kg
Calcium Sulfate, Dihydrate, NF is used as an excipient for its filler and diluent properties. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
Quantity 50 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.