Alfa Aesar™ Calcium sulfide, 99.9% (metals basis excluding Sr), Sr typically 500 ppm

Manufacturer: Alfa Aesar™ 1312106

Quantity 5g

 View more versions of this product

Catalog No. AA1312106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20548-54-3
    Formula Weight 72.14
    MDL Number MFCD00015990
    UN Number UN3134
    Merck Index 14,1707
    Chemical Name or Material Calcium sulfide

    Specifications

    Formula Weight 72.14
    Health Hazard 3 GHS P Statement
    P231+P232-P261-P280-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −325 Mesh
    Health Hazard 2 GHS H Statement
    H261-H315-H319-H335
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00015990
    Merck Index 14,1707
    Health Hazard 1 Warning
    Density 2.5g/mL
    Physical Form Powder
    Sensitivity Moisture Sensitive
    CAS 20548-54-3
    UN Number UN3134
    Odor Stench
    Quantity 5g
    DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.
    Chemical Name or Material Calcium sulfide
    Assay 99.9%, (metals basis excluding Sr), Sr typically 500ppm

    GHS H Statement
    H261-H315-H319-H335
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P231+P232-P261-P280-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit