Learn More
Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%
Description
Calcium sulfide is used in phosphors, lubricant additives and luminous paints. It reacts with elemental sulfur to give a lime-sulfur. Further, it is used in ore dressing and flotation agent. In addition to this, it is used as a depilatory in the tanning industry and in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN3134 |
Merck Index | 14,1707 |
Solubility Information | Slightly soluble in water. Insoluble in alcohol. |
Formula Weight | 72.14 |
Percent Purity | 99% |
Odor | Stench |
Show More |
Safety and Handling
Hazard Category | H228-H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H261-H315-H319-H335 In contact with water releases flammable gas. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.1; Packaging Group: II |
EINECSNumber | 243-873-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.