Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium tetrafluoroborate hydrate

Catalog No. AA3013122
Click to view available options
Quantity:
25 g
100 g

CAS: 13814-93-2 | B2CaF8 | 213.685 g/mol

Calcium tetrafluoroborate hydrate is used as pharmaceutical and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13814-93-2
Molecular Formula B2CaF8
Molecular Weight (g/mol) 213.685
MDL Number MFCD00015972
InChI Key YHLNCCJTKRTQEG-UHFFFAOYSA-N
Synonym calcium tetrafluoroborate, calcium ditetrafluoroborate, calcium tetrafluoroborate 1-, acmc-1bo7f
PubChem CID 6096284
IUPAC Name calcium;ditetrafluoroborate
SMILES [B-](F)(F)(F)F.[B-](F)(F)(F)F.[Ca+2]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water.
Formula Weight 213.68 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Calcium tetrafluoroborate hydrate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 237-485-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.