Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium thioglycolate trihydrate, 99%

Catalog No. p-2691162
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 65208-41-5 | C2H10CaO5S | 186.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65208-41-5
Molecular Formula C2H10CaO5S
Molecular Weight (g/mol) 186.24
MDL Number MFCD00150707
InChI Key GDECCIJZCWLFSB-UHFFFAOYSA-N
Synonym calciumthioglycolatetrihydrate
PubChem CID 131881455
SMILES O.O.O.[Ca].OC(=O)CS

Specifications

Melting Point 270°C to 280°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 1713
Solubility Information Solubility in water: 70g/L (25°C). Other solubilities: very slightly soluble in alcohol,chloroform,practical insoluble in ether,petroleum ether,,benzene
IUPAC Name calcium;2-sulfanylacetic acid;trihydrate
Formula Weight 184.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Calcium thioglycolate trihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.