Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium thiosulfate, pure, 30-50% solution in water, Acros Organics

Click to view available options
Quantity:
1L
5L
10124-41-1,7732-18-5
CaO3S2
152.21
FAYYUXPSKDFLEC-UHFFFAOYSA-L
calcium thiosulfate,tecesal,calcium thiosulphate,calcium hyposulfite,caswell no. 153,unii-2io10o728e,calcium thiosulfate ca s2o3,epa pesticide chemical code 080101,thiosulfuric acid h2s2o3 , calcium salt 1:1,thiosulfuric acid, calcium salt 1:1
24964
calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
[O-]S(=O)(=S)[O-].[Ca+2]
10124-41-1,7732-18-5
CaO3S2
152.21
FAYYUXPSKDFLEC-UHFFFAOYSA-L
calcium thiosulfate,tecesal,calcium thiosulphate,calcium hyposulfite,caswell no. 153,unii-2io10o728e,calcium thiosulfate ca s2o3,epa pesticide chemical code 080101,thiosulfuric acid h2s2o3 , calcium salt 1:1,thiosulfuric acid, calcium salt 1:1
24964
calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
[O-]S(=O)(=S)[O-].[Ca+2]
Specific Gravity 1.245,1.245
Linear Formula CaS2O3
Solubility Information Solubility in water: soluble
Formula Weight 152.21
Grade Pure
Density 1.2450g/mL
Quantity 5L
Chemical Name or Material Calcium thiosulfate

EINECSNumber : 233-333-7